Skip to Content
Merck
All Photos(1)

Key Documents

97085

Supelco

Benzylidene camphor sulfonic acid

analytical standard

Synonym(s):

α-(2-Oxoborn-3-ylidene)-toluene-4-sulphonic acid, 3-Benzylidenecamphor-4′-sulphonic acid, 4′-Sulfo-3-benzylidenecamphor, 4-[(4,7,7-Trimethyl-3-oxobicyclo[2.2.1]hept-2-ylidene)methyl]benzenesulfonic acid, p-[(2-Oxo-3-bornylidene)methyl]benzenesulfonic acid, BCSA

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€154.00

€154.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€154.00

About This Item

Empirical Formula (Hill Notation):
C17H20O4S
CAS Number:
Molecular Weight:
320.40
Beilstein:
14648450
UNSPSC Code:
85151701
NACRES:
NA.24

€154.00


Estimated to ship on21 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤8.0% water (calc. from elemental analysis)

application(s)

environmental

format

neat

SMILES string

CC1(C)[C@@H]2CC[C@]1(C)C(/C2=C\C3=CC=C(S(=O)(O)=O)C=C3)=O

InChI

1S/C17H20O4S/c1-16(2)14-8-9-17(16,3)15(18)13(14)10-11-4-6-12(7-5-11)22(19,20)21/h4-7,10,14H,8-9H2,1-3H3,(H,19,20,21)/b13-10-

InChI key

KKJKXQYVUVWWJP-RAXLEYEMSA-N

General description

Benzylidene camphor sulfonic acid (BCSA) is an organic ultraviolet absorbing compound used primarily in cosmetics (sunscreens, lipsticks, shampoos and hair sprays).[1]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic UV filters and their photodegradates, metabolites and disinfection by-products in the aquatic environment.
Diaz-Cruz MS, et al.
TrAC, Trends in Analytical Chemistry, 27(10), 873-887 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service