Skip to Content
Merck
All Photos(2)

Key Documents

96365

Supelco

Ponceau 6R

analytical standard

Synonym(s):

Crystal Ponceau 6R, Acid Red 44, Brilliant Crystal Scarlet, Crystal Scarlet, Ponceau 6R

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€158.00

€158.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€158.00

About This Item

Empirical Formula (Hill Notation):
C20H12N2Na2O7S2
CAS Number:
Molecular Weight:
502.43
Colour Index Number:
16250
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E126
NACRES:
NA.24

€158.00


Estimated to ship on14 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].Oc1ccc2cc(cc(c2c1\N=N\c3cccc4ccccc34)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C20H14N2O7S2.2Na/c23-17-9-8-13-10-14(30(24,25)26)11-18(31(27,28)29)19(13)20(17)22-21-16-7-3-5-12-4-1-2-6-15(12)16;;/h1-11,23H,(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b22-21+;;

InChI key

FUGCXLNGEHFIOA-ZPZFBZIMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ponceau 6R is a red azo dye which is widely used as a food colorant and also used in histology staining. It is soluble in water and slightly soluble in ethanol.[1][2]

Application

Ponceau 6R (P6R) has been used:
  • As a reference standard for the determination of synthetic food and beverages colorants by high-performance liquid chromatography coupled with diode-array detector.[3]
  • As a model compound for detailed study of the mechanism of an azo-dye degradation by soybean peroxidase enzyme (SBP).[4]
It may be used as a reference standard in the determination of ponceau 6R in drinks and candy samples using reversed-phase high performance liquid chromatography (RP-HPLC).[5]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinghua Hu et al.
Journal of molecular and cellular cardiology, 139, 113-123 (2020-01-27)
Transient receptor potential canonical 1 (TRPC1) protein is abundantly expressed in cardiomyocytes. While TRPC1 is supposed to be critically involved in cardiac hypertrophy, its physiological role in cardiomyocytes is poorly understood. We investigated the subcellular location of TRPC1 and its
Luchao Qi et al.
IEEE transactions on bio-medical engineering, 67(4), 1074-1082 (2019-07-25)
Invasiveness research is an essential step in breast cancer metastasis. The application of high-frequency ultrasound microbeam stimulation (HFUMS) offers a manner of determining the invasion potential of human breast cancer cells by eliciting the elevation of transient cytoplasmic calcium ions
Synthetic Dyes in Biology, Medicine And Chemistry (2012)
Soybean peroxidase-mediated degradation of an azo dye?a detailed mechanistic study.
Ali L, et al.
BMC Biochemistry, 14(1), 35-35 (2013)
Li-Ying Zhang et al.
Breast cancer research and treatment, 182(1), 21-33 (2020-05-18)
Previous studies have indicated that transient receptor potential (TRP) channels can influence cancer development. The TRPC subfamily consists of seven subtypes, TRPC1 - TRPC7. Interestingly, the expression levels of TRPC1 have been shown to be totally different in different breast cancer cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service