Skip to Content
Merck
All Photos(1)

Key Documents

96175

Sigma-Aldrich

3-tert-Butyl-4-hydroxyanisole

tested according to Ph. Eur.

Synonym(s):

Butylhydroxyanisolum, 2-tert-Butyl-4-methoxyphenol, 3-BHA, BHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O2
CAS Number:
Molecular Weight:
180.24
Beilstein:
1867499
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

solid

application(s)

pharmaceutical (small molecule)

SMILES string

COc1ccc(O)c(c1)C(C)(C)C

InChI

1S/C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12/h5-7,12H,1-4H3

InChI key

MRBKEAMVRSLQPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Matsuoka et al.
Mutation research, 241(2), 125-132 (1990-06-01)
The mutagenicity of 3-tert-butyl-4-hydroxyanisole (BHA) and its metabolites was investigated in the reverse mutation assay using S. typhimurium strains and the chromosomal aberration test in vitro using a Chinese hamster fibroblast cell line, CHL. BHA, tert-butylhydroquinone (BHQ), tert-butylquinone (BQ) and
K Tajima et al.
Drug metabolism and disposition: the biological fate of chemicals, 20(6), 816-820 (1992-11-01)
3-Tert-butyl-4-hydroxyanisole (3-BHA) was metabolized in the presence of horseradish peroxidase and hydrogen peroxide to 2-tert-butyl-p-benzoquinone (TBQ), 2,3-epoxy-5-tert-butyl-1,4-benzoquinone (TBQ-epoxide), and two known dimers. The formation of TBQ from 3-BHA required both horseradish peroxidase and hydrogen peroxide. When 2.5 mM 3-BHA was
L K Lam et al.
Carcinogenesis, 12(12), 2311-2315 (1991-12-01)
Carcinogen-induced aberrant crypts (AC) of the colon are a precancerous state that leads to malignancy. The inhibition of AC formation by chemopreventive agents was evaluated in this study. Colon AC were induced by 1,2-dimethylhydrazine (DMH) in 3 weeks in CF1
M G Salgo et al.
Archives of biochemistry and biophysics, 333(2), 482-488 (1996-09-15)
Peroxynitrite is a strong oxidant that reacts with a variety of biomolecules in vivo and in vitro. When rat thymocytes in phosphate buffer are exposed to 25 microM peroxynitrite for 10 min, DNA single strand breaks (SSB) can be detected.
K Morimoto et al.
Carcinogenesis, 12(4), 703-708 (1991-04-01)
DNA damage in the forestomach epithelium of male F344 rats was tested by alkaline elution assay following oral administration of 3-tert-butyl-4-hydroxyanisole (3-BHA) and its free metabolites. Although 1% 3-BHA and 0.001% tert-butylhydroquinone (BHQ) caused no detectable DNA damage, a tert-butylquinone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service