Skip to Content
Merck
All Photos(1)

Key Documents

94907

Sigma-Aldrich

4-Vinylbenzyl chloride

technical, ≥90% (GC), contains 0.1% total stabilizer

Synonym(s):

4-(Chloromethyl)styrene

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€126.00
250 ML
€434.00

€126.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€126.00
250 ML
€434.00

About This Item

Linear Formula:
H2C=CHC6H4CH2Cl
CAS Number:
Molecular Weight:
152.62
Beilstein:
2204384
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€126.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

technical

Quality Level

Assay

≥90% (GC)

form

liquid

contains

0.05% 4-tert-butylcatechol as stabilizer
0.05% nitroparaffin as stabilizer
0.1% total stabilizer

refractive index

n20/D 1.572 (lit.)
n20/D 1.572

bp

229 °C (lit.)

density

1.083 g/mL at 20 °C
1.083 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCc1ccc(C=C)cc1

InChI

1S/C9H9Cl/c1-2-8-3-5-9(7-10)6-4-8/h2-6H,1,7H2

InChI key

ZRZHXNCATOYMJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Influence of functional group concentration on hypercrosslinking of poly (vinylbenzyl chloride) PolyHIPEs: The research investigates how the concentration of 4-vinylbenzyl chloride affects the hypercrosslinking and properties of PolyHIPEs, enhancing their macroporosity (Koler et al., 2021).
  • Optimization of Synthesis of Poly (vinylbenzyl chloride) by RAFT Polymerisation: This study optimizes the RAFT polymerization conditions for synthesizing poly (vinylbenzyl chloride), analyzing the effect of different isomers in the reaction (Allozy & Abd Karim, 2021).
  • Effect of styrene addition on chemically induced grafting of 4-vinylbenzyl chloride onto low-density polyethylene: This paper explores the effects of styrene addition on the graft polymerization of 4-vinylbenzyl chloride onto polyethylene, aiming to improve the material′s properties for use in anion exchange membranes (Chen et al., 2019).
  • Synthesis and characterization of potentially polymerizable amine-derived ionic liquids bearing 4-vinylbenzyl group: The research focuses on synthesizing new amine-derived ionic liquids with 4-vinylbenzyl groups, evaluating their polymerizability and potential applications in materials science (Zajac et al., 2019).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tien D Ho et al.
Analytica chimica acta, 843, 18-26 (2014-08-26)
Super elastic nitinol (NiTi) wires were exploited as highly robust supports for three distinct crosslinked polymeric ionic liquid (PIL)-based coatings in solid-phase microextraction (SPME). The oxidation of NiTi wires in a boiling (30%w/w) H2O2 solution and subsequent derivatization in vinyltrimethoxysilane

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service