Skip to Content
Merck
All Photos(1)

Key Documents

94735

Sigma-Aldrich

Vanadyl acetylacetonate

purum, ≥97.0% (RT)

Synonym(s):

VO(acac)2, Vanadium(IV)-oxy acetylacetonate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€47.80

€47.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€47.80

About This Item

Linear Formula:
OV(C5H7O2)2
CAS Number:
Molecular Weight:
265.16
Beilstein:
4138236
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€47.80


Check Cart for Availability

Request a Bulk Order

grade

purum

Quality Level

Assay

≥97.0% (RT)

form

solid

reaction suitability

reagent type: catalyst

mp

235 °C (dec.) (lit.)

SMILES string

CC(=O)\C=C(\C)O[V](=O)O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.O.V/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;;/q;;;+2/p-2/b2*4-3-;;

InChI key

JFHJZWAQYMGNBE-SUKNRPLKSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marvin W Makinen et al.
The Journal of biological chemistry, 277(14), 12215-12220 (2002-01-30)
We have investigated the interaction of bis(acetylacetonato)oxovanadium(IV) (VO(acac)(2)) with bovine serum albumin (BSA) by EPR and angle-selected electron nuclear double resonance, correlating results with assays of glucose uptake by 3T3-L1 adipocytes. EPR spectra of VO(acac)(2) showed no broadening in the
[Oxidation of phenothiazine and its N-methyl derivative by the combination of iodobenzene and vanadium acetylacetonate].
F Pautet et al.
Die Pharmazie, 40(3), 202-203 (1985-03-01)
N Farrell et al.
Journal of inorganic biochemistry, 30(4), 309-315 (1987-08-01)
A study of the reactions of purine and pyrimidine bases has been initiated with the V(IV) complex [V(O)(acac)2]. Two distinct modes of reaction have been recognized depending on the nature of the base. Simple adduct formation occurs when the base

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service