Skip to Content
Merck
All Photos(1)

Key Documents

94355

Supelco

3-Hydroxyisovaleryl-L-carnitine-(N-methyl-d3)

analytical standard

Synonym(s):

(2R)-3-Carboxy-2-(3-hydroxy-3-methyl-1-oxobutoxy)-N,N-dimethyl-N-(methyl-d3)-1-propanaminium inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D3H20NO5
CAS Number:
Molecular Weight:
264.33
MDL number:
UNSPSC Code:
41116107
Product 94355 is currently not available for purchase. Contact Technical Service

grade

analytical standard

Quality Level

isotopic purity

≥99.0 atom % D

Assay

≥90.0% (HPLC)

optical activity

[α]/D -16.0±2.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

C[N+](C)(C([2H])([2H])[2H])C[C@H](OC(CC(C)(O)C)=O)CC([O-])=O

InChI

1S/C12H23NO5/c1-12(2,17)7-11(16)18-9(6-10(14)15)8-13(3,4)5/h9,17H,6-8H2,1-5H3/t9-/m1/s1/i3D3

InChI key

IGLHHSKNBDXCEY-MCHJGEEVSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service