Skip to Content
Merck
All Photos(2)

Key Documents

94319

Supelco

Omeprazole sulfone

analytical standard

Synonym(s):

5-Methoxy-2-{[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl}-1H-benzimidazole, Omeprazole sulphone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€294.00

€294.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€294.00

About This Item

Empirical Formula (Hill Notation):
C17H19N3O4S
CAS Number:
Molecular Weight:
361.42
Beilstein:
8347309
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€294.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=S(C1=NC2=CC(OC)=CC=C2N1)(CC3=NC=C(C)C(OC)=C3C)=O

InChI

1S/C17H19N3O4S/c1-10-8-18-15(11(2)16(10)24-4)9-25(21,22)17-19-13-6-5-12(23-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)

InChI key

IXEQEYRTSRFZEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shabnam Nazir et al.
European journal of drug metabolism and pharmacokinetics, 40(1), 13-22 (2014-01-07)
Omeprazole (OMP) is effective in the treatment of gastric hyperacidity and is metabolized by CYP2C19 and CYP3A4. These enzymes are modulated by estrogen and progesterone which regulate the menstrual cycle. The variations in the pharmacokinetics (PK) of many drugs like

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service