Skip to Content
Merck
All Photos(1)

Documents

94180

Sigma-Aldrich

10-Undecenoyl chloride

technical, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)8COCl
CAS Number:
Molecular Weight:
202.72
Beilstein:
1635112
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥97.0% (GC)

refractive index

n20/D 1.454 (lit.)
n20/D 1.454

bp

120-122 °C/10 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)CCCCCCCCC=C

InChI

1S/C11H19ClO/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2H,1,3-10H2

InChI key

MZFGYVZYLMNXGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service