Skip to Content
Merck
All Photos(1)

Key Documents

93874

Supelco

Fluroxypyr

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-[(4-Amino-3,5-dichloro-6-fluoro-2-pyridinyl)oxy]acetic acid, 4-Amino-3,5-dichloro-6-fluoro-2-pyridyloxyacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€86.50

€86.50


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
€86.50

About This Item

Empirical Formula (Hill Notation):
C7H5Cl2FN2O3
CAS Number:
Molecular Weight:
255.03
Beilstein:
7136185
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€86.50


Estimated to ship on17 May 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

Nc1c(Cl)c(F)nc(OCC(O)=O)c1Cl

InChI

1S/C7H5Cl2FN2O3/c8-3-5(11)4(9)7(12-6(3)10)15-1-2(13)14/h1H2,(H2,11,12)(H,13,14)

InChI key

MEFQWPUMEMWTJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Fluroxypyr is a systemic herbicide that belongs to the pyridine family and is widely used in crop fields for controlling the post-emergence broadleaf weeds. Fluroxypyr is applied in ester form and after uptake by the leaf, it is hydrolyzed to fluroxypyr acid, which is translocated rapidly to other parts of plants. It induces auxin-like responses, disrupts the key metabolic process, and causes lethal symptoms in plants

The Acceptable Daily Intake (ADI) and an Acceptable Operator Exposure Level (AOEL) for Fluroxypyr has been defined at 0.8 mg/kg body weight. Fluroxypyr is approved for use in European Union (EU) as per Regulation (EC) No 1107/2009. The maximum residue level (MRLs) for Fluroxypyr (sum of fluroxypyr, its salts, esters, and conjugates, expressed as fluroxypyr) has been set from 0.01 mg/kg to 2 mg/kg for products of plants and animal origin following Regulation (EU) 2021/1098.

Fluroxypyr has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The Fluroxypyr CRM may also find following uses:
  • Evaluate the eco-toxicity of fluroxypyr to green algae, bioaccumulation, and degradation of fluroxypyr in Chlamydomonas reinhardtii, followed by investigation of its biological adaptation to fluroxypyr toxicity
  • Study the effects of soil microbe, soil type, dissolved organic matter, temperature, soil moisture, and surfactant on fluroxypyr degradation in soils
  • Study the photocatalytic degradation of herbicide fluroxypyr in aqueous suspension of TiO2
  • Investigate the adsorption and thermal desorption of the herbicide fluroxypyr on activated carbon fibers and cloth at different pH values
  • Study the hydrolysis, aqueous photolysis, and soil degradation of fluroxypyr under laboratory conditions

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service