Skip to Content
Merck
All Photos(1)

Key Documents

93513

Supelco

4-Hexylresorcinol

analytical standard

Synonym(s):

4-Hexyl-1,3-dihydroxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€72.60

€72.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€72.60

About This Item

Linear Formula:
CH3(CH2)5C6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
194.27
Beilstein:
2048312
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€72.60


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

bp

333-335 °C (lit.)

mp

65-67 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCCCc1ccc(O)cc1O

InChI

1S/C12H18O2/c1-2-3-4-5-6-10-7-8-11(13)9-12(10)14/h7-9,13-14H,2-6H2,1H3

InChI key

WFJIVOKAWHGMBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hexylresorcinol is a well-known food additive belonging to the class of dihydroxybenzenes.[1] As an alternative to sulfite, 4-hexylresorcinol is proposed for use as a processing aid for the prevention of melanosis in shrimp and crab, which occurs due to the polyphenol oxidase activity during storage.[1][2][3]

Application

4-Hexylresorcinol may be used as an analytical reference standard for the quantification of the analyte in food matrices[1] and deepwater pink shrimp[3] using chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

388.4 °F

Flash Point(C)

198 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of an analytical method for the determination of 4-hexylresorcinol in food
Kim H-Y, et al.
Food Chemistry, 190, 1086-1092 (2016)
Melanosis inhibition and 4-hexylresorcinol residual levels in deepwater pink shrimp (Parapenaeus longirostris) following various treatments
Montero P, et al.
European Food Research and Technology, 223(1), 16-16 (2006)
Generally recognized as safe (GRAS) evaluation of 4-hexylresorcinol for use as a processing aid for prevention of melanosis in shrimp
Frankos HV, et al.
Regulatory Toxicology and Pharmacology, 14(2), 202-212 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service