Skip to Content
Merck

Skip To

93035

Malonyl-L-carnitine-(N-methyl-d3) lithium salt

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 466,00

About This Item

Empirical Formula (Hill Notation):
C10D3H14NO6 · xLi+
CAS Number:
Molecular Weight:
250.26 (free acid basis)
UNSPSC Code:
41116107
NACRES:
NA.24

€ 466,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥92.0% (HPLC)

form

solid

optical activity

[α]/D -28.0±2.0°, c = 0.1 in H218O

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99.0%

impurities

2.0-3.0% lithium

application(s)

clinical testing

storage temp.

−20°C

SMILES string

C[N+](C[C@H](OC(CC(O)=O)=O)CC([O-])=O)(C)C([2H])([2H])[2H]

InChI

1S/C10H17NO6/c1-11(2,3)6-7(4-8(12)13)17-10(16)5-9(14)15/h7H,4-6H2,1-3H3,(H-,12,13,14,15)/t7-/m1/s1/i1D3

InChI key

ZGNBLKBZJBJFDG-ISLPBHIQSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
926619258615868
grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

assay

≥92.0% (HPLC)

assay

≥95.0% (HPLC)

assay

≥95.0% (HPLC)

assay

≥93.0% (HPLC)

extent of labeling

≥99.0%

extent of labeling

≥99.0%

extent of labeling

≥99.0%

extent of labeling

-

optical activity

[α]/D -28.0±2.0°, c = 0.1 in H218O

optical activity

[α]/D -15.0±2.0°, c = 0.1 in H2O

optical activity

[α]/D -17.0±2.0°, c = 0.1 in H2O

optical activity

[α]0.1/D -18.0±2.0° in H2O

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters