Skip to Content
Merck
All Photos(1)

Documents

92951

Supelco

Vasicine

analytical standard

Synonym(s):

(−)-Peganine, (3S)-1,2,3,9-Tetrahydropyrrolo[2,1-b]quinazolin-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O
CAS Number:
Molecular Weight:
188.23
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

O[C@@H]1C2=NC3=CC=CC=C3CN2CC1

InChI

1S/C11H12N2O/c14-10-5-6-13-7-8-3-1-2-4-9(8)12-11(10)13/h1-4,10,14H,5-7H2/t10-/m0/s1

InChI key

YIICVSCAKJMMDJ-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chronic toxicity studies with vasicine from Adhatoda vasica Nees. in rats and monkeys.
G S Pahwa et al.
Indian journal of experimental biology, 25(7), 467-470 (1987-07-01)
Deepak Kumar Jha et al.
Applied biochemistry and biotechnology, 168(5), 980-990 (2012-08-18)
The extraction and determination of alkaloids was performed and confirmed by phytochemical analysis. Six different quinazoline alkaloids (vasicoline, vasicolinone, vasicinone, vasicine, adhatodine and anisotine) were found in the leaf of Justicia adhatoda (J. adhatoda). The presence of the peaks obtained
Pharmacokinetics of vasicine in healthy Indian volunteers.
V Amla et al.
Zhongguo yao li xue bao = Acta pharmacologica Sinica, 8(2), 190-192 (1987-03-01)
Bin Liu
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(11), 1719-1721 (2012-04-18)
To study the chemical consituents of Peganum multisectum. The compounds were isolated by chromatographic methods and their structures were elucidated by physicochemical properties and spectral analysis. Six alkaloids were isolated and identifiedas deoxyvasicinone (1), l-vasicinone (2), harmine (3), vasicine (4)
Bharat Singh et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 20(5), 441-445 (2013-01-30)
Adhatoda vasica Nees, Acanthaceae, is well known plant in Ayurveda and Unani medicine. The purpose of this study was to characterize the most bioactive phytochemicals viz., vasicine, vasicinone, vasicine acetate, 2-acetyl benzyl amine, vasicinolone present in the chloroform fraction having

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service