Skip to Content
Merck
All Photos(2)

Key Documents

92842

Supelco

Pyributicarb

PESTANAL®, analytical standard

Synonym(s):

O-3-tert-Butylphenyl 6-methoxy-2-pyridyl(methyl)thiocarbamate, O-[3-(1,1-Dimethylethyl)phenyl] N-(6-methoxy-2-pyridinyl)-N-methylcarbamothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22N2O2S
CAS Number:
Molecular Weight:
330.44
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 64.1-66.7%
nitrogen content, 8.3-8.7%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cccc(n1)N(C)C(=S)Oc2cccc(c2)C(C)(C)C

InChI

1S/C18H22N2O2S/c1-18(2,3)13-8-6-9-14(12-13)22-17(23)20(4)15-10-7-11-16(19-15)21-5/h6-12H,1-5H3

InChI key

VTRWMTJQBQJKQH-UHFFFAOYSA-N

General description

Pyributicarb is a systemic, thiocarbamate herbicide, which is effectively used for the post-emergence control of annual grass weeds, perennial grasses, turf, etc. Its mode of action involves inhibition of elongation of roots and aerial parts in weeds.[1]

Application

Pyributicarb may be used as an analytical reference standard for the quantification of the analyte in agricultural products using gas chromatography technique.[2][3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multi-residue analysis of 205 crop pesticides using mini-solid phase extraction?large volume injection?GC?MS
Xu L-X, et al.
Journal of Pharmaceutical and Biomedical Analysis, 70(1-2), 173-183 (2009)
The Pesticide Encyclopedia (2014)
Validation of multiresidue screening methods for the determination of 186 pesticides in 11 agricultural products using gas chromatography (GC).
Hirahara Y, et al.
Journal of Health Science, 51(5), 617-627 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service