Skip to Content
Merck
All Photos(1)

Key Documents

92822

Sigma-Aldrich

Picrylsulfonic acid solution

1 M in H2O

Synonym(s):

2,4,6-Trinitrobenzenesulfonic acid solution, TNBS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3N3O9S
CAS Number:
Molecular Weight:
293.17
Beilstein:
572358
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.25

form

liquid

Quality Level

concentration

1 M in H2O

density

1.14 g/mL at 20 °C

ε (extinction coefficient)

≤0.85 at 340 nm

shipped in

dry ice

storage temp.

−20°C

SMILES string

OS(=O)(=O)c1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3N3O9S/c10-7(11)3-1-4(8(12)13)6(19(16,17)18)5(2-3)9(14)15/h1-2H,(H,16,17,18)

InChI key

NHJVRSWLHSJWIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrophilic modifying reagent. Derivatives with amino compounds can be regenerated with hydrazine.
Picrylsulfonic acid (2,4,6-Trinitrobenzenesulfonic acid solution, TNBS) may be used in studies on the mechanisms of intracellular pH regulation. TNBS is used to induce inflammatory bowel disease (IBD)/colonic inflammation for studies on its mechanism and treatment.

Caution

decomposes when stored at temperatures ≥0°C with precipitate formation (picric acid)

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Desen. Expl. 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Beatriz Albino de Almeida et al.
Journal of ethnopharmacology, 146(1), 300-310 (2013-01-15)
In Brazilian traditional medicine, Arctium lappa (Asteraceae), has been reported to relieve gastrointestinal symptoms. In the present study, we investigated the effects of the lactone sesquiterpene onopordopicrin enriched fraction (ONP fraction) from Arctium lappa in an experimental colitis model induced
Joel Castro et al.
Gastroenterology, 145(6), 1334-1346 (2013-08-21)
Linaclotide is a minimally absorbed agonist of guanylate cyclase-C (GUCY2C or GC-C) that reduces symptoms associated with irritable bowel syndrome with constipation (IBS-C). Little is known about the mechanism by which linaclotide reduces abdominal pain in patients with IBS-C. We
A Arranz et al.
Journal of controlled release : official journal of the Controlled Release Society, 165(3), 163-172 (2012-11-28)
CD40-CD40L interactions appear to play an important role in the pathogenesis of experimental colitis. We tested the effect and investigated the underlying mechanism of action of systemically administered antisense oligonucleotide (ASO) targeting CD40 formulated in amphoteric liposomes (nov038/CD40). The charge
Zhen Huang et al.
Gut, 63(8), 1247-1257 (2013-09-04)
Emerging evidence suggests that microRNA (miRNA)-mediated gene regulation influences a variety of autoimmune disease processes, including Crohn's disease (CD), but the biological function of miRNAs in CD remains unclear. We examine miRNA level in colon tissues and study the potential
Bas Kremer et al.
PloS one, 7(11), e50388-e50388 (2012-12-12)
The recurrent TNBS-colitis model in BALB/c mice has been proposed as a model of Inflammatory Bowel Disease with a shifting pattern of local cytokines with the expression of Th1 cytokines during the early phase, Th17 cytokines during the intermediate phase

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service