Skip to Content
Merck
All Photos(1)

Key Documents

92650

Millipore

Strep B ChromoSelect Selective Supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€31.00
5 VIALS
€140.00

€31.00


Estimated to ship on19 April 2025


Request more information

Select a Size

Change View
1 VIAL
€31.00
5 VIALS
€140.00

About This Item

UNSPSC Code:
41171614
NACRES:
NB.21

€31.00


Estimated to ship on19 April 2025


Request more information

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Streptococcus spp.

Application

An antibiotic supplement recommended for the selective isolation of Group B Streptococci from clinical samples.

Components

Composition:
(per vial sufficient for 1000 ml medium)
Colistin 10mg
Nalidixic acid 10mg
Gentamicin 2mg

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service