Skip to Content
Merck

Skip To

92571

mono-Methyl fumarate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Fumaric acid monomethyl ester, Methyl hydrogen fumarate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 181,00

About This Item

Linear Formula:
HO2CCH=CHCO2CH3
CAS Number:
Molecular Weight:
130.10
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 181,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

shelf life

1440 days

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

mp

144-145 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)\C=C\C(O)=O

InChI

1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2+

InChI key

NKHAVTQWNUWKEO-NSCUHMNNSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
471477294516967
technique(s)

gas chromatography (GC): suitable

technique(s)

gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

-

application(s)

environmental
pharmaceutical

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under mono-Methyl fumarate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters