Skip to Content
Merck
All Photos(2)

Documents

92549

Supelco

Acenaphthylene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8
CAS Number:
Molecular Weight:
152.19
Beilstein:
774092
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

280 °C (lit.)

mp

78-82 °C (lit.)
89-92 °C

density

0.899 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

c1cc2C=Cc3cccc(c1)c23

InChI

1S/C12H8/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-8H

InChI key

HXGDTGSAIMULJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

251.6 °F - closed cup

Flash Point(C)

122.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Onruthai Pinyakong et al.
FEMS microbiology letters, 238(2), 297-305 (2004-09-11)
Sphingomonas sp. strain A4 is capable of utilizing acenaphthene and acenaphthylene as sole carbon and energy sources, but it is unable to grow on other polycyclic aromatic hydrocarbons (PAHs). The genes encoding terminal oxygenase components of ring-hydroxylating dioxygenase (arhA1 and
Zhiru Ma et al.
The journal of physical chemistry. A, 111(10), 2020-2027 (2007-03-29)
13C chemical shift tensor data from 2D FIREMAT spectra are reported for 4,7-di-t-butylacenaphthene and 4,7-di-t-butylacenaphthylene. In addition, calculations of the chemical shielding tensors were completed at the B3LYP/6-311G** level of theory. While the experimental tensor data on 4,7-di-t-butylacenaphthylene are in
N D Marsh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56A(8), 1499-1511 (2000-07-25)
We have examined the UV spectra of six newly-synthesized ethynyl-PAH (1-ethynylnaphthalene, 5-ethynylacenaphthylene, 1-ethynylanthracene, 9-ethynylphenanthrene, 3-ethynylfluoranthene, and 1-ethynylpyrene) and five corresponding CP-PAH (acenaphthylene, aceanthrylene, acephenanthrylene, cyclopenta[cd]fluoranthene, and cyclopenta[cd]pyrene) and have found the following systematic behavior: ethynyl-substitution results in average bathochromic shifts
Anand S Nayak et al.
Biodegradation, 20(6), 837-843 (2009-06-23)
Stenotrophomonas sp. RMSK capable of degrading acenaphthylene as a sole source of carbon and energy was isolated from coal sample. Metabolites produced were analyzed and characterized by TLC, HPLC and mass spectrometry. Identification of naphthalene-1,8-dicarboxylic acid, 1-naphthoic acid, 1,2-dihydroxynaphthalene, salicylate
E Leonhardt et al.
Analytical chemistry, 70(6), 1228-1230 (1998-04-08)
Polycyclic aromatic hydrocarbons were extracted from a soil sample using ultrasound and dichloromethane-, cyclohexane-, and toluene-water mixtures. It was found that when dichloromethane is used as an extractant, acenaphthylene reacts with the solvent. Several chlorinated and oxygenated derivatives were identified.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service