Skip to Content
Merck
All Photos(1)

Key Documents

92413

Supelco

D-(+)-Melibiose

analytical standard

Synonym(s):

6-O-α-D-Galactopyranosyl-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93360
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

optical activity

[α]/D 137.0±3.0°, 10 hr, c = 5 in H2O

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

oligosaccharides

impurities

≤7.0% water

application(s)

food and beverages

format

neat

SMILES string

OC[C@H]1O[C@H](OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H22O11/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12/h1,4-12,14-21H,2-3H2/t4-,5+,6+,7+,8+,9-,10-,11+,12-/m0/s1

InChI key

AYRXSINWFIIFAE-GFRRCQKTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Melibiose is a disaccharide,[1] composed of galactose and glucose with an α-1,6 glycoside linkage. It is reported to promote the absorption of quercetin glycosides in the ligated small intestinal loop of rats.[2]

Application

D-(+)-Melibiose may be used as an analytical reference standard for the determination of the analyte in soybean samples using normal-phase high-performance liquid chromatography (HPLC) with evaporative light scattering detection[3] and in environmental samples by gas chromatography-mass spectrometry (GC-MS) technique.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple analytical method for high-throughput screening of major sugars from soybean by normal-phase HPLC with evaporative light scattering detection
Valliyodan B, et al.
Chromatography Research International, 2015 (2015)
Analysis of sugars in environmental samples by gas chromatography?mass spectrometry
Medeiros MP, et al.
Journal of Chromatography A, 1141(2), 271-278 (2007)
Melibiose, a nondigestible disaccharide, promotes absorption of quercetin glycosides in rat small intestine
Tanaka S, et al.
Journal of Agricultural and Food Chemistry, 64(49), 9335-9341 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service