Skip to Content
Merck
All Photos(1)

Key Documents

92264

Supelco

4-Methoxyphenol

analytical standard

Synonym(s):

4-Hydroxyanisole, 4-MP, HQMME, Hydroquinone monomethyl ether, MEHQ, p-Guaiacol

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€51.50

€51.50


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€51.50

About This Item

Linear Formula:
CH3OC6H4OH
CAS Number:
Molecular Weight:
124.14
Beilstein:
507924
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€51.50


Estimated to ship on25 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

4.3 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥98.5% (GC)

autoignition temp.

789 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

bp

243 °C (lit.)

mp

55-57 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1ccc(O)cc1

InChI

1S/C7H8O2/c1-9-7-4-2-6(8)3-5-7/h2-5,8H,1H3

InChI key

NWVVVBRKAWDGAB-UHFFFAOYSA-N

Gene Information

human ... TYR(7299)

Looking for similar products? Visit Product Comparison Guide

General description

4-Methoxyphenol belongs to the class of hydroquinone (HQ) derivatives. It is widely used as a skin whitening agent, because it can exert hypo-pigmenting effects.[1] It also finds applications as a stabilizer for chlorinated hydrocarbons, ethyl cellulose, UV (ultraviolet) inhibitor, inhibitor for acrylic monomers, etc.[2]

Application

4-Methoxyphenol may be used as an analytical reference standard for the quantification of the analyte in cosmetic products[1][3] using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proctor and Hughes' Chemical Hazards of the Workplace, 50(5-6), 346-352 (2004)
Experimental and statistical validation of HPLC analysis of hydroquinone and its 4-methoxyphenol, 4-ethoxyphenol and 4-benzyloxyphenol ethers in cosmetic products
Borremans M, et al.
Chromatographia, 50(5-6), 346-352 (1999)
Quantitative determination of $\alpha$-arbutin, $\beta$-arbutin, kojic acid, nicotinamide, hydroquinone, resorcinol, 4-methoxyphenol, 4-ethoxyphenol, and ascorbic acid from skin whitening products by HPLC-UV
Wang YH, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 98(1), 5-12 (2015)
Travis M Doggett et al.
Alcoholism, clinical and experimental research, 38(9), 2414-2426 (2014-09-27)
Alcohol intoxication can increase inflammation and worsen injury, yet the mechanisms involved are not clear. We investigated whether acute alcohol intoxication increases microvascular permeability and investigated potential signaling mechanisms in endothelial cells that may be involved. Conscious rats received a
Chongming Liu et al.
Journal of mass spectrometry : JMS, 49(8), 692-699 (2014-07-22)
On contrary to the widely accepted conviction that the m/z 93 ion derived from phenol does not react with CO2, we demonstrate that it makes an adduct with CO2 to a small but demonstrable extent. For example, the product-ion mass

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service