Skip to Content
Merck
All Photos(1)

Key Documents

92075

Sigma-Aldrich

1,3,5-Triisopropylbenzene

purum, ≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3[CH(CH3)2]3
CAS Number:
Molecular Weight:
204.35
Beilstein:
1862750
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥95.0% (GC)

refractive index

n20/D 1.488 (lit.)
n20/D 1.488

bp

232-236 °C (lit.)

density

0.845 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1cc(cc(c1)C(C)C)C(C)C

InChI

1S/C15H24/c1-10(2)13-7-14(11(3)4)9-15(8-13)12(5)6/h7-12H,1-6H3

InChI key

VUMCUSHVMYIRMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selvedin Telalović et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(7), 2077-2088 (2011-01-25)
Bimetallic three-dimensional amorphous mesoporous materials, Al-Zr-TUD-1 materials, were synthesised by using a surfactant-free, one-pot procedure employing triethanolamine (TEA) as a complexing reagent. The amount of aluminium and zirconium was varied in order to study the effect of these metals on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service