Skip to Content
Merck
All Photos(1)

Key Documents

91556

Supelco

3-Benzylidenecamphor

analytical standard

Synonym(s):

1,7,7-Trimethyl-3-(phenylmethylene)bicyclo[2.2.1]heptan-2-one, 3-BC, 3-Benzylidene-1,7,7-trimethyl-norbornan-2-one, 3-Benzylidene-2-bornanone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€87.90

€87.90


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€87.90

About This Item

Empirical Formula (Hill Notation):
C17H20O
CAS Number:
Molecular Weight:
240.34
Beilstein:
1967440
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€87.90


Estimated to ship on12 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

C[C@]1(C/2=O)CC[C@@H](C1(C)C)C2=C\C3=CC=CC=C3

InChI

1S/C17H20O/c1-16(2)14-9-10-17(16,3)15(18)13(14)11-12-7-5-4-6-8-12/h4-8,11,14H,9-10H2,1-3H3/b13-11-

InChI key

OIQXFRANQVWXJF-QBFSEMIESA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elissa O'Malley et al.
The Science of the total environment, 754, 142373-142373 (2020-12-02)
UV filters present in sunscreen and other cosmetics are directly released into the environment during aquatic recreational activities. The extent to which the wide range of UV filters pose a risk to the environment remains unclear. This study investigated the
Rory Verhagen et al.
Chemosphere, 223, 731-737 (2019-02-27)
In recent years, organic ultraviolet filters (UVFs) received considerable attention as a group of emerging contaminants, including in Australia where the use of UVFs is particularly relevant. Passive sampling using polymers has become widely used for routine monitoring of chemicals
Adela Jing Li et al.
Journal of hazardous materials, 337, 115-125 (2017-05-17)
Ethyl-4-aminobenzoate (Et-PABA) is currently used as a substitute for 4-aminobenzoate (PABA) in sunscreens and anesthetic ointments. Despite its widespread use and hydrophilicity, Et-PABA has never been found in environmental waters. This study, probed the occurrence of Et-PABA in both seawater
Hanne Frederiksen et al.
International journal of hygiene and environmental health, 220(2 Pt A), 244-253 (2016-09-18)
Experimental studies indicate that some chemicals with UV blocking properties (known as UV filters) can act as endocrine disruptors. UV filters are used in sunscreens and other cosmetic- and personal care products, as well as in other consumer products such

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service