Skip to Content
Merck
All Photos(2)

Key Documents

908797

Sigma-Aldrich

HS161

Umicore

Synonym(s):

Trimethyl(methylcyclopentadienyl)platinum(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H4CH3Pt(CH3)3
CAS Number:
Molecular Weight:
319.30
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

crystals
low-melting solid

mp

29-30 °C
30-31 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Pt](C)C.C[C]1[CH][CH][CH][CH]1

InChI

1S/C6H7.3CH3.Pt/c1-6-4-2-3-5-6;;;;/h2,4H,3H2,1H3;3*1H3;

InChI key

AGQKROMSWCHOND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Umicore HS161 is a homogeneous catalyst useful for hydrosilylation reactions.

Storage and Stability

Do not store above 4°C (39°F). Store in a tightly closed original container under dry inert gas protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service