All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H25FOSi
CAS Number:
Molecular Weight:
232.41
UNSPSC Code:
12352101
Recommended Products
Assay
≥95%
form
liquid
storage temp.
2-8°C
Application
The incorporation of fluorine in agrochemical and pharmaceutical compounds is of high interest due to the ability to fine-tune the polarity, pKA-value, conformation and metabolic stability. Derived from a cis-selective hydrogenation developed by Glorius and coworkers, this fluorinated compound displays a diverse utility as a building block for chemical synthesis. Fluoro-cyclohexanol allows for the straightforward application in organic synthesis, after simple deprotection. It can be utilized to prepare fluorinated analogues of known compounds.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mario P Wiesenfeldt et al.
Science (New York, N.Y.), 357(6354), 908-912 (2017-08-12)
All-cis-multifluorinated cycloalkanes exhibit intriguing electronic properties. In particular, they display extremely high dipole moments perpendicular to the aliphatic ring, making them highly desired motifs in material science. Very few such motifs have been prepared, as their syntheses require multistep sequences
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service