Skip to Content
Merck
All Photos(1)

Key Documents

90221

Supelco

Retigabine

analytical standard

Synonym(s):

D-23129, Ezogabine, N-(2-Amino-4-(4-fluorobenzylamino)phenyl)carbamic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€115.00

€115.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€115.00

About This Item

Empirical Formula (Hill Notation):
C16H18FN3O2
CAS Number:
Molecular Weight:
303.33
Beilstein:
8072099
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€115.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

drug control

USDEA Schedule IV

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water (calc. from elemental analysis)

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

FC1=CC=C(CNC2=CC(N)=C(NC(OCC)=O)C=C2)C=C1

InChI

1S/C16H18FN3O2/c1-2-22-16(21)20-15-8-7-13(9-14(15)18)19-10-11-3-5-12(17)6-4-11/h3-9,19H,2,10,18H2,1H3,(H,20,21)

InChI key

PCOBBVZJEWWZFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Retigabine is an antiepileptic drug, which activates neuronal KCNQ-type K+ channels by inducing a large hyperpolarizing shift of steady-state activation.[1][2]

Application

Retigabine may be used as a reference standard in the determination of retigabine in biological matrices using high-performance liquid chromatography coupled with tandem mass spectrometry with positive ion atmospheric pressure chemical ionization (HPLC-APCI-MS/MS).[3]

Biochem/physiol Actions

Retigabine (Ezogabine) is a Kv7.2-7.5 (KCNQ2-5) neuronal potassium channel opener witrh anticonvulsant activity.
Retigabine is a Kv7.2-7.5 potassium channel activator; anticonvulsant.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The new anticonvulsant retigabine favors voltage-dependent opening of the Kv7. 2 (KCNQ2) channel by binding to its activation gate.
Wuttke VT, et al.
Molecular Pharmacology, 94, 71-76 (2005)
Determination of retigabine and its acetyl metabolite in biological matrices by on-line solid-phase extraction (column switching) liquid chromatography with tandem mass spectrometry.
Knebel GN, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 748(1), 97-111 (2000)
Identification, characterization, synthesis and HPLC quantification of new process-related impurities and degradation products in retigabine.
Dousa M, et al.
Journal of Pharmaceutical and Biomedical Analysis, 94, 71-76 (2014)
Lyubov I Brueggemann et al.
Molecular pharmacology, 86(3), 330-341 (2014-06-20)
Recent research suggests that smooth muscle cells express Kv7.4 and Kv7.5 voltage-activated potassium channels, which contribute to maintenance of their resting membrane voltage. New pharmacologic activators of Kv7 channels, ML213 (N-mesitybicyclo[2.2.1]heptane-2-carboxamide) and ICA-069673 N-(6-chloropyridin-3-yl)-3,4-difluorobenzamide), have been reported to discriminate among

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service