Skip to Content
Merck

Skip To

89983

Benzyl isothiocyanate

analytical standard

Synonym(s):

Benzyl mustard oil, Isothiocyanotaomethylbenzene

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
C6H5CH2NCS
CAS Number:
Molecular Weight:
149.21
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
210-753-9
Beilstein/REAXYS Number:
386135
MDL number:


grade

analytical standard

Quality Segment

assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.601 (lit.), n20/D 1.602-1.605

bp

242-243 °C (lit.)

density

1.125 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

S=C=NCc1ccccc1

InChI

1S/C8H7NS/c10-7-9-6-8-4-2-1-3-5-8/h1-5H,6H2

InChI key

MDKCFLQDBWCQCV-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


Still not finding the right product?

Explore all of our products under Benzyl isothiocyanate


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

222.8 °F

flash_point_c

106 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters