Skip to Content
Merck

89902

Sigma-Aldrich

p-Toluic acid

purum, ≥98.0% (GC)

Synonym(s):

4-Methylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CO2H
CAS Number:
Molecular Weight:
136.15
Beilstein:
507600
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Pricing and availability is not currently available.

grade

purum

Assay

≥98.0% (GC)

bp

274-275 °C (lit.)

mp

177-180 °C (lit.)
177-182 °C

SMILES string

Cc1ccc(cc1)C(O)=O

InChI

1S/C8H8O2/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

LPNBBFKOUUSUDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
U2625AG974286842
Uric acid ≥99%, crystalline

U2625

Uric acid

mp

30-34 °C (lit.)

mp

>300 °C (lit.)

mp

-

mp

60-61 °C (lit.)

bp

330 °C (lit.)

bp

-

bp

-

bp

-

form

crystalline, waxy solid, liquid

form

crystalline

form

powder

form

-

density

0.786 g/mL at 25 °C (lit.)

density

-

density

-

density

-

vapor density

9.27 (vs air)

vapor density

-

vapor density

-

vapor density

-

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

357.8 °F

Flash Point(C)

181 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sven Lahme et al.
Applied and environmental microbiology, 78(5), 1606-1610 (2011-12-20)
A novel alphaproteobacterium isolated from freshwater sediments, strain pMbN1, degrades 4-methylbenzoate to CO(2) under nitrate-reducing conditions. While strain pMbN1 utilizes several benzoate derivatives and other polar aromatic compounds, it cannot degrade p-xylene or other hydrocarbons. Based on 16S rRNA gene
C E Kuo et al.
Chemosphere, 45(6-7), 835-842 (2001-11-07)
The composition of the headspace gas affected the growth dynamics of microbial populations and the biotransformation pattern of p-toluic acid in anoxic estuarine sediments. Under CO2 atmosphere, p-toluic acid was transformed by the sediment microorganisms without a lag period, while
Sven Lahme et al.
Environmental microbiology, 14(5), 1118-1132 (2012-01-24)
The pathway for anaerobic degradation of 4-methylbenzoate was studied in the denitrifying alphaproteobacterium Magnetospirillum sp. strain pMbN1. Adaptation studies with whole cells indicated substrate-dependent induction of the capacity to degrade 4-methylbenzoate. Differential protein profiling (2D-DIGE) of 4-methylbenzoate- in comparison with
F Junker et al.
Microbiology (Reading, England), 142 ( Pt 9), 2419-2427 (1996-09-01)
Three multicomponent oxygenases involved in the degradation of p-toluenesulfonate and p-toluenecarboxylate and the regulation of their synthesis have been examined in three strains (T-2, PSB-4 and TER-1) of Comamonas testosteroni. Strain T-2 utilizes p-toluenesulfonate as a source of carbon and
A C Franck-Mokross et al.
Applied microbiology and biotechnology, 50(2), 233-240 (1998-12-05)
Pseudomonas sp. D7-4 and Pseudomonas sp. B13 FR1(pFRC20P) degraded mixtures of chloro- and methyl-substituted benzoates exclusively via an extended ortho pathway, whereas in Pseudomonas putida WR 201 both ortho and meta fission were induced by mixtures of 3-chloro- and 3-methylbenzoate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service