Skip to Content
Merck
All Photos(1)

Key Documents

89610

Sigma-Aldrich

o-Toluidine

purum p.a., ≥99.5% (GC)

Synonym(s):

2-Aminotoluene, 2-Methylaniline

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€63.30
500 ML
€101.00

€63.30


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€63.30
500 ML
€101.00

About This Item

Linear Formula:
CH3C6H4NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
741981
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€63.30


Estimated to ship on02 June 2025


Request a Bulk Order

vapor density

3.7 (vs air)

Quality Level

vapor pressure

0.26 mmHg ( 25 °C)

grade

purum p.a.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

899 °F

impurities

≤0.1% water
≤0.5% m- and p-toluidine (GC)

ign. residue

≤0.01% (as SO4)

refractive index

n20/D 1.572 (lit.)
n20/D 1.572

bp

199-200 °C (lit.)
89-90 °C/11 mmHg (lit.)

mp

−28 °C (lit.)

solubility

H2O: slightly soluble

density

1.008 g/mL at 25 °C (lit.)

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

SMILES string

Cc1ccccc1N

InChI

1S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5H,8H2,1H3

InChI key

RNVCVTLRINQCPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Toluidine belongs to the class of monocyclic aromatic amines, which are of research and industrial importance. It can be used as a derivatization agent for the resolution of analytes by the formation of chromophores, which can be visualized by UV radiation or fluorescence.[1][2]

Application

o-Toluidine may be used as a TLC Visualization (Staining) Reagent for the detection of reducing sugars in protein hydrolysates by using high-performance thin-layer chromatography (HPTLC) coupled with UV diffuse reflectance scanning densitometry.[1]

Caution

may discolor on storage

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of aromatic amines in textile after bromination, by gas chromatography coupled with electron capture detection
Dados AE, et al.
Chromatographia, 59(5-6), 335-341 (2004)
An o-toluidine method for detection of carbohydrates in protein hydrolysates
Morcol T and Velander WH
Analytical Biochemistry, 195(1), 153-159 (1991)
Tania Carreón et al.
Occupational and environmental medicine, 67(5), 348-350 (2009-11-04)
In 1991, the US National Institute for Occupational Safety and Health (NIOSH) reported an increased bladder cancer risk in a cohort of 1749 workers potentially exposed to o-toluidine and aniline at a chemical manufacturing plant. As additional information showed that
Hun-Je Jo et al.
Chemosphere, 70(6), 949-957 (2007-09-26)
In this study, major toxicants in pigment wastewaters (i.e. raw wastewater and effluent) were determined by toxicity identification evaluations (TIEs), and changes in their toxicities due to combined gamma-ray (gamma-ray) and ozone treatments were evaluated. From TIE results using Daphnia
Dirk Lindner et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 20(7), 1524-1536 (2011-05-27)
This multicenter, observational study was conducted in three European countries (Germany, Switzerland, and the United Kingdom) to determine the exposure of adult cigarette smokers and nonsmokers to selected cigarette smoke constituents: 1,3-butadiene, 2-naphthylamine, 4-aminobiphenyl, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK), acrolein, benzene, carbon monoxide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service