Skip to Content
Merck
All Photos(1)

Key Documents

87709

Supelco

Tetramethylammonium bromide

for electrochemical analysis, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(Br)
CAS Number:
Molecular Weight:
154.05
Beilstein:
3620955
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

Assay

≥99.0% (AT)
≥99.0%

form

crystals

mp

>300 °C (lit.)

solubility

H2O: 0.1 g/mL, clear, colorless

SMILES string

[Br-].C[N+](C)(C)C

InChI

1S/C4H12N.BrH/c1-5(2,3)4;/h1-4H3;1H/q+1;/p-1

InChI key

DDFYFBUWEBINLX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Matvienko et al.
PloS one, 8(2), e55913-e55913 (2013-02-15)
Several applications of high throughput genome and transcriptome sequencing would benefit from a reduction of the high-copy-number sequences in the libraries being sequenced and analyzed, particularly when applied to species with large genomes. We adapted and analyzed the consequences of
Konrad Rykaczewski et al.
Langmuir : the ACS journal of surfaces and colloids, 29(3), 881-891 (2012-12-25)
The prospect of enhancing the condensation rate by decreasing the maximum drop departure diameter significantly below the capillary length through spontaneous drop motion has generated significant interest in condensation on superhydrophobic surfaces (SHS). The mobile coalescence leading to spontaneous drop
Yusuke Ide et al.
Chemical communications (Cambridge, England), 48(56), 7073-7075 (2012-06-12)
Hiroshima University Silicate-1 (HUS-1), composed of silicate sheets with a halved sodalite cage and the interlayer tetramethylammonium (TMA) cation in the cage, was modified with dimethyldichlorosilane to form the organic derivative in which a dimethyl group was grafted onto the
Hui Chong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(4), 2091-2098 (2011-11-08)
A new water-soluble conjugated polymer containing fluorene and boron-dipyrromethene repeat units in the backbones (PBF) that exhibits red emission was synthesized and characterized. Cationic PBF forms uniform nanoparticles with negatively charged disodium salt 3,3'-dithiodipropionic acid (SDPA) in aqueous solution through
Yasuyuki Ishida et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 27(10), 1053-1056 (2011-10-12)
Thermochemolysis-gas chromatography in the presence of tetramethylammonium acetate was applied to the direct determination of terephthalic acid (TPA) contained in solid decomposition products obtained from the hydrothermal recycling process of poly(ethylene terephthalate) (PET). On the chromatograms of the hydrothermal decomposition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service