Skip to Content
Merck
All Photos(1)

Documents

86877

Supelco

Tetrabutylammonium methanesulfonate

≥97.0% (T)

Synonym(s):

Methanesulfonic acid tetrabutylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3CH2CH2CH2]4N(CH3SO3)
CAS Number:
Molecular Weight:
337.56
Beilstein:
4059876
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (T)

form

crystals

mp

78-80 °C

storage temp.

2-8°C

SMILES string

CS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.CH4O3S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

XHYYGJAYKIYARQ-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiantian Mou et al.
Scientific reports, 10(1), 18685-18685 (2020-10-31)
A translocator protein 18 kDa targeted radiotracer, N,N-diethyl-2-(2-(4-[18F]fluorophenyl)-5,7-dimethylpyrazolo[1,5-a] pyrimidin-3-yl) acetamide ([18F]FDPA), was automated synthetized and evaluated for cardiac inflammation imaging. Various reaction conditions for an automated synthesis were systematically optimized. MicroPET/CT imaging were performed on normal rats and rats with myocardial
Wouter Monnens et al.
Physical chemistry chemical physics : PCCP, 22(42), 24526-24534 (2020-10-23)
The electrochemical behavior and electrodeposition of indium was investigated at 26 °C and 160 °C from a solution composed of indium(iii) methanesulfonate and dimethylsulfoxide (DMSO). Indium(iii) methanesulfonate was synthesized from indium(iii) oxide and methanesulfonic acid (MSA). Cyclic voltammetry, quartz crystal
Shuverthi De Sarkar et al.
The European physical journal. E, Soft matter, 43(8), 55-55 (2020-08-21)
The physical properties of an aqueous solution of a macromolecule primarily depend on its chemical structure and the mesoscopic aggregates formed by many of such molecules. Ionic liquids (ILs) are the macromolecules that have caught significant research interests for their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service