Skip to Content
Merck

85702

Stearamide

technical, ~90% (GC)

Synonym(s):

Amide C18

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)16CONH2
CAS Number:
Molecular Weight:
283.49
EC Number:
204-693-2
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
909006
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

technical

assay

~90% (GC)

bp

250-251 °C/12 mmHg (lit.)

mp

98-102 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(N)=O

InChI

1S/C18H37NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H2,19,20)

InChI key

LYRFLYHAGKPMFH-UHFFFAOYSA-N

Other Notes

This product has been replaced by O601-ALDRICH | Octadecanamide technical grade, 85%




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library