Skip to Content
Merck
All Photos(1)

Key Documents

83920

Millipore

Rogosa Agar

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€183.00

€183.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€183.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

€183.00


Estimated to ship on28 May 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
ammonium citrate, 2 g/L
casein peptone, 10 g/L
ferrous sulfate, 0.034 g/L
D(+)-glucose, 20 g/L
magnesium sulfate, 0.575 g/L
manganous sulfate, 0.12 g/L
potassium dihydrogen phosphate, 6 g/L
sodium acetate, 15 g/L
yeast extract, 5 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

application(s)

bioburden testing
clinical testing
environmental
food and beverages

microbiology

suitability

selective for Bifidobacterium spp.
selective for Lactobacillus spp.

Application

Selective agar for the isolation and enumeration of lactobacilli belonging to the buccal and intestinal flora, as well as from food such as milk and meat.

Preparation Note

73 g are added to 1 litre distilled water and brought to the boil to dissolve. 10 ml sterile 10% Tween® 80 solution is added. The medium is adjusted to pH 5.5 with 1.32 ml glacial acetic acid. Fill into aseptic containers. Do NOT autoclave.

Other Notes

Selective media for the isolation and enumeration of lactobacilli[1]

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.E. Sharpe
Laboratory Practice, 9, 223-223 (1960)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service