Skip to Content
Merck
All Photos(1)

Documents

83551

Millipore

Rice Extract Agar

suitable for microbiology

Synonym(s):

REA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

sterility

non-sterile

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 14.3 g/L
rice extract concentrate, 0.7 g/L

final pH

5.8±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

nonselective for Candida spp.
nonselective for Saccharomyces spp.
nonselective for Zygosaccharomyces spp.

Application

Rice Extract Agar is a test medium acc. to Taschdjian for the differentiation of yeasts, in particular Candida albicans and Candida stellatoidea alongside other Candida species, based on their typical chlamydospores. Likewise, other species of yeast can be differentiated, on the basis of micromorphological criteria, with rice extract agar.

Other Notes

In mycological diagnostics

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald M. Atlas
Handbook of Microbiological Media null
L Sigler et al.
Journal of medical and veterinary mycology : bi-monthly publication of the International Society for Human and Animal Mycology, 32(4), 275-285 (1994-01-01)
Non-dermatophytic fungi are increasingly being recognized as agents of onychomycosis. In 1990, three cases of chronic infection of the great toenail in adult female residents of Canada were attributed to Onychocola canadensis, a previously unknown hyphomycete. Three additional cases were
B Willinger et al.
Mycoses, 37(11-12), 401-403 (1994-11-01)
The purpose of this study was to determine whether yeast isolates from mothers and their new-borns are of the same genotype. In this investigation, 103 parturient mothers and their children were examined for colonization by yeasts by sampling the vaginal

Articles

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service