Skip to Content
Merck
All Photos(1)

Key Documents

83461

Sigma-Aldrich

RBS 35 solution

concentrate

Synonym(s):

RBS 35 concentrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€110.00
5 L
€410.00
10 L
Inquire

€110.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€110.00
5 L
€410.00
10 L
Inquire

About This Item

UNSPSC Code:
41103206
NACRES:
NA.22

€110.00


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

form

liquid

impurities

≤1% sodium hydroxide

pH

11.4-12.5(at 2% v/v in water)(lit.)
12.9-13.9(pure)(lit.)

density

1.10 g/mL at 20 °C

General description

RBS 35 is a multipurpose alkaline detergent with high wetting and emulsifying properties that is suitable for use in hard water.[1] It can be used for manual cleaning, soaking or in ultrasonic baths.

Application

RBS 35 solution is used to clean glassware, microscope lenses and mirrors, and laboratory equipment made of stainless steel, plastic, porcelain, etc. This cleaning agent is also used for the removal of radioactive contamination.

Features and Benefits

  • Efficient at low concentrations
  • Alternative to solvents and sulfochromic acids
  • No risk of calcium/magnesium deposits when diluted in tap water
  • Biodegradable surfactant

Other Notes

Sales restrictions may apply

Legal Information

RBS is a trademark of Chemical Products R. Borghgraef S.A.

related product

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-adherence potential of Enterococcus durans cells and its cell-free supernatant on plastic and stainless steel against foodborne pathogens
Ait Meddour A, et al.
Folia Microbiologica, 60(4), 357-363 (2015)
Quantitative detection of Listeria monocytogenes in biofilms by real-time PCR
Morgan Guilbaud, et al.
Applied and Environmental Microbiology, 71(4), 2190-2194 (2005)

Questions

1–2 of 2 Questions  
  1. What is the working dilution of RBS™ 35 solution detergent (ref. 83461-5L)?

    1 answer
    1. The recommended use concentration for RBS™ 35 solution detergent (ref. 83461-5L) is 2% v/v in water.

      Helpful?

  2. ¿Cuál es la dilución de trabajo del detergente RBS™ 35 solution (ref. 83461-5L)? Actualmente aplicamos una dilución de 20ml detergente/1.000ml Agua Purificada. Gracias.

    1 answer
    1. The supplier recommends a working concentration of 2% v/v in water. Diluting 20 mL of this product in 1000 mL is appropriate.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service