Skip to Content
Merck
All Photos(1)

Key Documents

80559

Supelco

L-α-Hydroxyglutaric acid-13C5 disodium salt

analytical standard

Synonym(s):

(S)-2-Hydroxypentanedioic acid-1,2,3,4,5-13C5 disodium salt, Disodium (S)-2-hydroxyglutarate-13C5

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€406.00

€406.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€406.00

About This Item

Empirical Formula (Hill Notation):
13C5H6Na2O5
Molecular Weight:
197.04
UNSPSC Code:
41116107
NACRES:
NB.21

€406.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

isotopic purity

≥98.0%

Assay

≥98.0% (GC)

optical activity

[α]/D -8.0±1.5°, c = 1 in 0.1 M NaOH

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

O=[13C](O[Na])[13C@@H](O)[13CH2][13CH2][13C](O[Na])=O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service