Skip to Content
Merck
All Photos(2)

Key Documents

80024

Supelco

Benzoin

analytical standard

Synonym(s):

α-Hydroxy-α-phenylacetophenone, α-Hydroxybenzyl phenyl ketone, 2-Hydroxy-2-phenylacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COC6H5
CAS Number:
Molecular Weight:
212.24
Beilstein:
391839
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

194 °C/12 mmHg (lit.)

mp

134-138 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC(c1ccccc1)C(=O)c2ccccc2

InChI

1S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15H

InChI key

ISAOCJYIOMOJEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoins are a class of 2-hydroxy ketones that are important subunits of several biologically active compounds.

Application

Benzoin has been used as a reference standard for the analysis of benzoin as an analyte protectant in fruit and vegetable samples by gas chromatography-mass spectrometry (GC-MS) operating in selected ion monitoring (SIM) mode. It may be used as a derivatizing agent for:
  • Guanidino compounds in serum prior to their determination by micellar electrokinetic capillary (MEKC) chromatography (MEKC).
  • Metformin in pharmaceutical preparations and urine prior to its determination by spectrophotometry.
  • Famotidine and ranitidine in drugs and biological samples prior to their determination using high performance liquid chromatography-diode array detection (HPLC-DAD).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Asymmetric benzoin reaction catalyzed by benzoylformate decarboxylase.
Demir AS, et al.
Tetrahedron Asymmetry, 10(24), 4769-4774 (1999)
Spectrophotometric determination of metformin in pharmaceutical preparations, serum and urine using benzoin as derivatizing reagent.
Alamgir M, et al.
J. Chem. Soc. Pak., 36(2), 344-349 (2014)
HPLC Determination of Metformin, Famotidine and Ranitidine by Derivatization with Benzoin from Drugs and Biological Samples.
Alamgir MYKM, et al.
Pharmaceutica Analytica Acta, 8(546), 2-2 (2017)
Determination of guanidino compounds in serum by micellar electrokinetic capillary chromatography using benzoin as the derivatizing reagent.
Kandhro AJ, et al.
Analytical Letters, 43(13), 2049-2060 (2010)
Evaluation of analyte protectants to improve gas chromatographic analysis of pesticides.
Anastassiades M, et al.
Journal of Chromatography A, 1015(1-2), 163-184 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service