Skip to Content
Merck
All Photos(1)

Documents

79525

Sigma-Aldrich

Triethyl phosphonoacetate

purum, ≥97.0% (GC)

Synonym(s):

(Diethoxyphosphinyl)acetic acid ethyl ester, Diethyl ethoxycarbonylmethylphosphonate, NSC 13898, NSC 16128, Triethyl carboxymethylphosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)CH2CO2C2H5
CAS Number:
Molecular Weight:
224.19
Beilstein:
1343714
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (GC)

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.431 (lit.)
n20/D 1.432

bp

142-145 °C/9 mmHg (lit.)

density

1.13 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CP(=O)(OCC)OCC

InChI

1S/C8H17O5P/c1-4-11-8(9)7-14(10,12-5-2)13-6-3/h4-7H2,1-3H3

InChI key

GGUBFICZYGKNTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reactant involved in:
  • Horner-Wadsworth-Emmons reactions
  • Intramolecular Heck-type cyclization and isomerizations
  • Tsuji-Trost type reactions
  • Intramolecular aryne-ene reactions
  • Synthesis of aldehydes

Other Notes

A Horner-Wittig reagent, review; Improved procedure for the C2 homologation of esters to α,β-unsaturated esters; Synthesis of α-(hydroxymethyl)acrylate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T. Minami et al.
Synthesis, 333-333 (1991)
J.M. Takacs et al.
Tetrahedron Letters, 27, 1257-1257 (1986)
J. Boutagy et al.
Chemical Reviews, 74, 87-87 (1974)
J. Villieras et al.
Organic Syntheses, 66, 220-220 (1988)
M Rodriguez et al.
International journal of peptide and protein research, 39(3), 273-277 (1992-03-01)
Racemic "carba" pseudo-dipeptide units such as Gly-psi(CH2-CH2)-D,L-Xaa were obtained either through the Horner-Emmons condensation of N-tert.-butyloxycarbonyl-beta alaninal with the appropriate substituted triethyl phosphonacetate, or from commercially available 3-carbethoxy-2-piperidone.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service