Skip to Content
Merck
All Photos(1)

Key Documents

78321

Supelco

Chlorpropham-(isopropyl-d7)

PESTANAL®, analytical standard

Synonym(s):

Chlorpropham-d7, Isopropyl-d7 N-(3-chlorophenyl)carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€147.00

€147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€147.00

About This Item

Empirical Formula (Hill Notation):
C10D7H5ClNO2
Molecular Weight:
220.70
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€147.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

Related Categories

General description

Chlorpropham-(isopropyl-d7) is an isotope-labeled analog of the phenylcarbamate pesticide chlorpropham,[1] wherein all the isopropyl protons are replaced by deuterium.

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Residue levels of chlorpropham in individual tubers and composite samples of postharvest-treated potatoes
Lentza-Rizos C and Balokas A
Food Chemistry, 49(2), 710-714 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service