Skip to Content
Merck
All Photos(2)

Key Documents

78118BC

Millipore

MVP ICON® Protective Case

for use with MVP ICON® instrument

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€50.00

€50.00


Estimated to ship on28 May 2025


Request more information

Select a Size

Change View
1 EACH
€50.00

About This Item

UNSPSC Code:
41116500

€50.00


Estimated to ship on28 May 2025


Request more information

product line

MVP ICON®

manufacturer/tradename

MVP ICON®

application(s)

food and beverages

compatibility

for use with MVP ICON® instrument

storage temp.

room temp

Legal Information

MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service