Skip to Content
Merck
All Photos(1)

Key Documents

76886

Sigma-Aldrich

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride complex

for fluorescence

Synonym(s):

Ru(ddp)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H48Cl2N6Ru
CAS Number:
Molecular Weight:
1169.17
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

for fluorescence

Assay

≥95% (HPCE)

form

powder

solubility

DMF: soluble
chloroform: soluble
ethanol: soluble
methanol: soluble

fluorescence

λex 455 nm; λem 610-630 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

SMILES string

Cl[Ru]Cl.c1ccc(cc1)-c2ccnc3c2ccc4c(ccnc34)-c5ccccc5.c6ccc(cc6)-c7ccnc8c7ccc9c(ccnc89)-c%10ccccc%10.c%11ccc(cc%11)-c%12ccnc%13c%12ccc%14c(ccnc%13%14)-c%15ccccc%15

InChI

1S/3C24H16N2.2ClH.Ru/c3*1-3-7-17(8-4-1)19-13-15-25-23-21(19)11-12-22-20(14-16-26-24(22)23)18-9-5-2-6-10-18;;;/h3*1-16H;2*1H;/q;;;;;+2/p-2

InChI key

SKZWFYFFTOHWQP-UHFFFAOYSA-L

Application

Ruthenium-tris(4,7-diphenyl-1,10-phenanthroline) dichloride (Ru(dpp)) has been widely used as a probe for luminescent detection and quantitation of oxygen. The fluorescence of the dye (abs. λmax 455 nm, luminescence λmax 613 nm) is strongly reduced by molecular oxygen due to dynamic quenching, making Ru(dpp) a viable oxygen probe based on either measurement of intensity or decay time. Ru(ddp) has been used for fiber optic sensors, studies on oxygen in skin and skin tumors, measurement of oxygen flux through skin, and for oxygen imaging.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W L Rumsey et al.
Science (New York, N.Y.), 241(4873), 1649-1651 (1988-09-23)
The imaging of phosphorescence provides a method for monitoring oxygen distribution within the vascular system of intact tissues. Isolated rat lives were perfused through the portal vein with media containing palladium coproporphyrin, which phosphoresced and was used to image the
G. Liebsch et al.
Appl. Spectroscopy, 54, 548-548 (2000)
Yuta Ando et al.
Advanced healthcare materials, 8(5), e1900001-e1900001 (2019-02-09)
Despite its revolutionary success in hematological malignancies, chimeric antigen receptor T (CAR-T) cell therapy faces disappointing clinical results in solid tumors. The poor efficacy has been partially attributed to the lack of understanding in how CAR-T cells function in a
Federica Tonon et al.
Scientific reports, 9(1), 13557-13557 (2019-09-21)
Among the multiple metabolic signals involved in the establishment of the hepatic zonation, oxygen could play a key role. Indeed, depending on hepatocyte position in the hepatic lobule, gene expression and metabolism are differently affected by the oxygen gradient present
Yuta Ando et al.
Scientific reports, 7(1), 15233-15233 (2017-11-11)
Hypoxia plays a central role in cancer progression and resistance to therapy. We have engineered a microdevice platform to recapitulate the intratumor oxygen gradients that drive the heterogeneous hypoxic landscapes in solid tumors. Our design features a "tumor section"-like culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service