Skip to Content
Merck
All Photos(1)

Key Documents

76734

Supelco

Daminozide-(dimethyl-d6) hydrochloride

PESTANAL®, analytical standard

Synonym(s):

Succinic acid mono[2,2-di(methyl-d3)hydrazide] hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€430.00

€430.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€430.00

About This Item

Empirical Formula (Hill Notation):
C6D6H6N2O3 · HCl
Molecular Weight:
202.67
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€430.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

OC(CCC(NN(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)=O.Cl

InChI key

NKMJPUKNMIGCFK-TXHXQZCNSA-N

General description

Daminozide-(dimethyl-d6) hydrochloride is an isotope-labeled analog of the plant growth regulator daminozide,[1] wherein all the dimethyl protons are replaced by deuterium.

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoulei Yan et al.
Biosensors & bioelectronics, 22(6), 1087-1091 (2006-04-20)
As the daminozide (DM) and its metabolite have been identified to be potentially carcinogenic, rapid detection method for them is necessary for food safety. A type of piezoelectric crystal sensor has been prepared by using a molecularly imprinted polymer (MIP)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service