Skip to Content
Merck
All Photos(1)

Key Documents

75825

Sigma-Aldrich

Sodium 2-oxobutyrate

≥99.0% (NT)

Synonym(s):

2-Oxobutanoic acid sodium salt, 2-Oxobutyric acid sodium salt, Sodium α-ketobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COCOONa
CAS Number:
Molecular Weight:
124.07
Beilstein:
3631701
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99.0% (NT)

SMILES string

[Na+].CCC(=O)C([O-])=O

InChI

1S/C4H6O3.Na/c1-2-3(5)4(6)7;/h2H2,1H3,(H,6,7);/q;+1/p-1

InChI key

SUAMAHKUSIHRMR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Substrate for the determination of lactate dehydrogenase isoenzymes.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service