Skip to Content
Merck
All Photos(1)

Key Documents

75602

Supelco

6-Prenylnaringenin

analytical standard

Synonym(s):

2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-6-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20O5
CAS Number:
Molecular Weight:
340.37
Beilstein:
5766535
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC1=C(C(CC(C2=CC=C(O)C=C2)O3)=O)C3=CC(O)=C1CC=C(C)C

InChI

1S/C20H20O5/c1-11(2)3-8-14-15(22)9-18-19(20(14)24)16(23)10-17(25-18)12-4-6-13(21)7-5-12/h3-7,9,17,21-22,24H,8,10H2,1-2H3/t17-/m0/s1

InChI key

YHWNASRGLKJRJJ-KRWDZBQOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: humulus
For this product we use the non-stereospecific CAS number. It is expected that the natural enantiomer is predominant, however the substance is prone to racemization in solution. The product is therefore not specified stereospecifically and is only recommended to be used for non-stereospecific analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carla Araya-Cloutier et al.
Scientific reports, 8(1), 9267-9267 (2018-06-20)
Prenylated flavonoids and isoflavonoids are phytochemicals with remarkable antibacterial activity. In this study, 30 prenylated (iso)flavonoids were tested against Listeria monocytogenes and Escherichia coli (the latter in combination with an efflux pump inhibitor). Minimum inhibitory concentrations of the most active
Tareisha L Dunlap et al.
Chemical research in toxicology, 30(11), 2084-2092 (2017-10-07)
Many women consider botanical dietary supplements (BDSs) as safe alternatives to hormone therapy for menopausal symptoms. However, the effect of BDSs on breast cancer risk is largely unknown. In the estrogen chemical carcinogenesis pathway, P450 1B1 metabolizes estrogens to 4-hydroxylated
Martin Ambrož et al.
Nutrients, 11(4) (2019-04-24)
Beer, the most popular beverage containing hops, is also frequently consumed by cancer patients. Moreover, non-alcoholic beer, owing to its nutritional value and high content of biological active compounds, is sometimes recommended to patients by oncologists. However, the potential benefits
Milou G M van de Schans et al.
Chembiochem : a European journal of chemical biology, 16(18), 2668-2677 (2015-10-10)
Six prenylated (iso)flavonoids were purified from a licorice root extract and subjected to competition experiments with six commercially available (iso)flavonoids. The agonistic and antagonistic activities of these compounds towards both hERα (human estrogen receptor alpha) and hERβ were determined. Differences
Ying-Yu Hsu et al.
Journal of food science and technology, 56(4), 1939-1953 (2019-04-19)
This study developed a high performance liquid chromatography with diode array detection (HPLC-DAD) and tandem mass spectrometry (MS-MS) method for determination of prenylflavonoids and hop bitter acids in surplus yeast, a byproduct from beer brewing process. This method enabled the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service