Skip to Content
Merck
All Photos(1)

Key Documents

74507

Sigma-Aldrich

cis-endo-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride

purum, ≥97.0% (GC)

Synonym(s):

cis-endo-5-Norbornene-2,3-dicarboxylic anhydride, Nadic anhydrid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O3
CAS Number:
Molecular Weight:
164.16
Beilstein:
83657
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

purum

Assay

≥97.0% (GC)

mp

164-165 °C (lit.)

SMILES string

O=C1OC(=O)[C@H]2[C@@H]3C[C@@H](C=C3)[C@@H]12

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malcolm Driffield et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(6), 1200-1213 (2018-02-24)
The polymeric coating used in metal packaging such as cans for foods and beverages may contain residual amounts of monomers used in the production of the coating, as well as unreacted linear and cyclic oligomers. Traditionally, although designed for use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service