Skip to Content
Merck
All Photos(2)

Documents

74412

Supelco

Trolamine salicylate

analytical standard

Synonym(s):

TEA salicylate, Triethanolamine salicylate, Triethanolaminium salicylate, Tris(2-hydroxyethyl)ammonium 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21NO6
CAS Number:
Molecular Weight:
287.31
Beilstein:
7451205
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

OCC[NH+](CCO)CCO.Oc1ccccc1C([O-])=O

InChI

1S/C7H6O3.C6H15NO3/c8-6-4-2-1-3-5(6)7(9)10;8-4-1-7(2-5-9)3-6-10/h1-4,8H,(H,9,10);8-10H,1-6H2

InChI key

UEVAMYPIMMOEFW-UHFFFAOYSA-N

General description

Trolamine salicylate is a type of salicylate derivative, which is a sunscreen agent and a topical analgesic.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C D Ciccone et al.
Physical therapy, 71(9), 666-675 (1991-09-01)
The purpose of this study was to determine the effects of ultrasound and phonophoresis using an anti-inflammatory-analgesic cream (trolamine salicylate) on delayed-onset muscle soreness (DOMS). Repeated eccentric contractions were used to induce DOMS in the elbow flexors of 40 college-aged
Brian G Ragan et al.
Medicine and science in sports and exercise, 39(11), 1942-1948 (2007-11-08)
Analgesic balms (AB) are widely used in sports medicine. We previously have examined effects of various counterirritant-based AB on pressor responses evoked by muscular contraction (MC), mediated by group III and IV muscle afferents known to produce exercise and nociceptive
Contact & Occupational Dermatology (2016)
J L Rabinowitz et al.
Journal of clinical pharmacology, 24(11-12), 532-539 (1984-11-01)
The transdermal absorption by tissues of salicylate from triethanolamine salicylate (TEA/S) ointment applied to canine and human skin was shown to be consistent, significant, and reproducible. Salicylate levels were measured using thin-layer chromatography, nuclear magnetic resonance, and radiochemistry. Absorption was
P Morra et al.
The Annals of pharmacotherapy, 30(9), 935-940 (1996-09-01)
To compare the rate and extent of systemic salicylate absorption following single and multiple applications of two topically applied analgesics, one containing methyl salicylate and the other containing trolamine salicylate. Two-period, two-treatment, randomized, crossover, multiple-dose study in healthy men and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service