Skip to Content
Merck
All Photos(1)

Key Documents

74103

Supelco

Triacetin

analytical standard

Synonym(s):

1,2,3-Triacetoxypropane, 1,2,3-Triacetylglycerol, Glyceryl triacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€203.00

€203.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€203.00

About This Item

Linear Formula:
(CH3COOCH2)2CHOCOCH3
CAS Number:
Molecular Weight:
218.20
Beilstein:
1792353
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€203.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

7.52 (vs air)

Assay

≥99.0% (GC)

autoignition temp.

809 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

1.05 %, 189 °F
7.73 %, 215 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.429-1.432
n25/D 1.429-1.431 (lit.)

bp

258 °C (lit.)

mp

3 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)OCC(COC(C)=O)OC(C)=O

InChI

1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3

InChI key

URAYPUMNDPQOKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monice Zondlo Fiume
International journal of toxicology, 22 Suppl 2, 1-10 (2003-10-14)
Triacetin, also known as Glyceryl Triacetate, is reported to function as a cosmetic biocide, plasticizer, and solvent in cosmetic formulations, at concentrations ranging from 0.8% to 4.0%. It is a commonly used carrier for flavors and fragrances. Triacetin was affirmed
Hywel D Williams et al.
Journal of pharmaceutical sciences, 103(6), 1734-1746 (2014-04-18)
The current study determined the extent to which the desorption of lipid-based formulations (LBFs) from a mesoporous magnesium aluminometasilicate (Neusilin®-US2) carrier is governed by drug properties, LBF composition, and LBF-to-adsorbent ratio. A secondary objective was to evaluate the impact of
Soon-Seok Hong et al.
International journal of pharmaceutics, 483(1-2), 142-150 (2015-02-11)
A high drug-loading capacity is a critical factor for the clinical development of liposomal formulations. The accommodation of hydrophobic drugs within the liposomal membrane is often limited in saturated phosphatidylcholine (PC)-based liposomes owing to the rigidity of the lipid acyl
Mohammad Sajid Ali et al.
Iranian journal of pharmaceutical research : IJPR, 13(4), 1125-1140 (2015-01-15)
Benign prostatic hyperplasia (BPH)is the most common condition in aging men, associated with lower urinary tract symptoms. It is caused due to the augmented levels of the androgen dihydrotestosterone. Dutasteride, a 5α-Reductase inhibitor has been recommended for the treatment of
Takayuki Sato et al.
Chemical & pharmaceutical bulletin, 63(2), 88-94 (2015-03-10)
The purpose of this study was to characterize the non-aqueous nanosuspension of a hydrophilic drug prepared by bead milling for cutaneous application. Riboflavin was used as the model hydrophilic drug. The non-aqueous nanosuspensions were prepared by grinding riboflavin with zirconia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service