Skip to Content
Merck
All Photos(1)

Documents

73906

Supelco

4,4′-Difluorobenzophenone

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(FC6H4)2CO
CAS Number:
Molecular Weight:
218.20
Beilstein:
516231
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

102-105 °C (lit.)

application(s)

environmental
pharmaceutical

format

neat

SMILES string

Fc1ccc(cc1)C(=O)c2ccc(F)cc2

InChI

1S/C13H8F2O/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8H

InChI key

LSQARZALBDFYQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Difluorobenzophenone is typically used as a starting material for the preparation of polyetheretherketone (PEEK) polymer, a high-temperature thermoplastic, which finds wide use in dentistry as well as in automotive, oil industries, etc.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

4,4′-Difluorobenzophenone (DFBP) may be used as an analytical reference standard for the determination of the analyte in water samples and DFBP/valerophenone mixture solutions by various chromatography techniques.

Analysis Note

The purity is calculated based on the principle of mass balance and is mentioned on the CoA

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F - Pensky-Martens closed cup

Flash Point(C)

152 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Applications of polyetheretherketone (PEEK) in oral implantology and prosthodontics
Najeeb S, et al.
Journal of Prosthodontic Research, 60(1), 12-19 (2016)
An improved automatic analyzer for organic compounds in water based on headspace solid-phase microextraction (SPME) coupled to gas chromatography
Belau E, et al.
Field Analytical Chemistry & Technology, 5(1-2), 37-49 (2001)
An automatic analyzer for organic compounds in water based on solid-phase microextraction coupled to gas chromatography
Grote C, et al.
Analytical Chemistry, 71(20), 4513-4518 (1999)
Intelligent peak deconvolution through in-depth study of the data matrix from liquid chromatography coupled with a photo-diode array detector applied to pharmaceutical analysis
Arase S, et al.
Journal of Chromatography A, 1469(1-2), 35-47 (2016)
The formation of 4, 4?-difluorobenzophenone from 4, 4?-dinitrodiphenylmethane
Adams DJ, et al.
Journal of Fluorine Chemistry, 92(2), 127-129 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service