Skip to Content
Merck
All Photos(1)

Key Documents

73747

Supelco

Phenethyl acetate

analytical standard

Synonym(s):

2-Phenethyl acetate, 2-Phenylethyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€203.00

€203.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€203.00

About This Item

Linear Formula:
CH3COOCH2CH2C6H5
CAS Number:
Molecular Weight:
164.20
Beilstein:
638179
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€203.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

5.67 (vs air)

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.498 (lit.)

bp

238-239 °C (lit.)

density

1.032 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)OCCc1ccccc1

InChI

1S/C10H12O2/c1-9(11)12-8-7-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3

InChI key

MDHYEMXUFSJLGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D McGinty et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S491-S497 (2012-03-15)
A toxicologic and dermatologic review of phenethyl acetate when used as a fragrance ingredient is presented. Phenethyl acetate is a member of the fragrance structural group aryl alkyl alcohol simple acid esters (AAASAE). The AAASAE fragrance ingredients are prepared by
Stéphanie Rollero et al.
Applied microbiology and biotechnology, 99(5), 2291-2304 (2014-11-22)
Volatile compounds produced by yeast during fermentation greatly influence the organoleptic qualities of wine. We developed a model to predict the combined effects of initial nitrogen and phytosterol content and fermentation temperature on the production of volatile compounds. We used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service