Skip to Content
Merck
All Photos(1)

Key Documents

73142

Millipore

Canavanine Glycine Bromothymol blue Agar

suitable for microbiology, NutriSelect® Basic

Synonym(s):

CGB Agar

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€294.00

€294.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€294.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

€294.00


Estimated to ship on17 May 2025


Request a Bulk Order

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 20.0 g/L
bromothymol blue, 0.08 g/L
L-canavanine sulfate, 0.03 g/L
glycine, 10.0 g/L
magnesium sulfate, 1.0 g/L
potassium phosphate, 1.0 g/L
thyamine hydrochloride, 0.001 g/L

manufacturer/tradename

NutriSelect® Basic

final pH

5.8±0.2 (25 °C)

application(s)

microbiology

storage temp.

2-8°C

Application

CGB Agar is recommended for the differentiation of Cryptococcus neoformans var. neoformans and Cryptococcus gattii

Preparation Note

Suspend 32.11 g in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Mix well and use as desired.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service