Skip to Content
Merck
All Photos(2)

Key Documents

73132

Supelco

meso-Erythritol

analytical standard

Synonym(s):

1,2,3,4-Butanetetrol, meso-1,2,3,4-Tetrahydroxybutane, i-Erythritol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CH(OH)]2CH2OH
CAS Number:
Molecular Weight:
122.12
Beilstein:
1719753
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

bp

329-331 °C (lit.)

mp

118-120 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

OC[C@@H](O)[C@@H](O)CO

InChI

1S/C4H10O4/c5-1-3(7)4(8)2-6/h3-8H,1-2H2/t3-,4+

InChI key

UNXHWFMMPAWVPI-ZXZARUISSA-N

Looking for similar products? Visit Product Comparison Guide

General description

meso-Erythritol, belonging to the class of sugar alcohols, is identified in a variety of food products, fruits, vegetables, beverages and dietary supplements.[1][2] It is known as a low glycemic food additive and plays an important role as a sweetener for diabetic patients, since it does not have glycemic or insulinemic effect due to its ability to not get metabolized but get absorbed in the small intestine.[3] It is also reportedly used as a sugar substitute in toothpaste, chewing gums, confectionery food products, etc.[4]

Application

meso-Erythritol may be used as an analytical reference standard for the quantification of the analyte in:
  • Food products using high-pressure liquid chromatography coupled to charged aerosol detector (HPLC-CAD).[1]
  • Confectioneries using high-performance liquid chromatography with UV detection (HPLC-UV).[2]
  • Food and beverages using liquid chromatography with UV detection (LC-UV).[4]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Allelic variation of the Tas1r3 gene affects behavioral taste responses to this sugar alcohol, suggesting that it is a T1R3 receptor ligand.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Liquid chromatographic determination of sugar alcohols in beverages and foods after nitrobenzoylation
Nojiri S, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 82(1), 134-140 (1999)
Zorn H and Czermak P
Biotechnology of Food and Feed Additives. (2014)
Zorn H and Czermak P
Biotechnology of Food and Feed Additives. (2014)
Analysis of erythritol in foods by polyclonal antibody-based indirect competitive ELISA
Sreenath K and Venkatesh PY
Analytical and Bioanalytical Chemistry, 391(2), 609-615 (2008)
Simultaneous separation and determination of erythritol, xylitol, sorbitol, mannitol, maltitol, fructose, glucose, sucrose and maltose in food products by high performance liquid chromatography coupled to charged aerosol detector
Grembecka M, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 117, 77-82 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service