Skip to Content
Merck
All Photos(1)

Key Documents

72994

Supelco

Neoxanthin

analytical standard

Synonym(s):

(3S,3′S,5R,5′R,6R,6′S,9′-cis)-6,7-Didehydro-5′,6′-epoxy-5′,6′-dihydro-β,β-carotene-3,3′,5(6H)-triol, 9′-cis-Neoxanthin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€694.00

€694.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€694.00

About This Item

Empirical Formula (Hill Notation):
C40H56O4
CAS Number:
Molecular Weight:
600.87
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

€694.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

C/C(C([H])=[C@]=C1[C@](C)(O)C[C@@H](O)CC1(C)C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(/C=C/[C@@]2(O3)[C@@]3(C)C[C@@H](O)CC2(C)C)C

InChI

1S/C40H56O4/c1-29(17-13-19-31(3)21-22-35-36(5,6)25-33(41)27-38(35,9)43)15-11-12-16-30(2)18-14-20-32(4)23-24-40-37(7,8)26-34(42)28-39(40,10)44-40/h11-21,23-24,33-34,41-43H,25-28H2,1-10H3/b12-11+,17-13+,18-14+,24-23+,29-15+,30-16+,31-19+,32-20+/t22?,33-,34-,38+,39+,40-/m0/s1

InChI key

PGYAYSRVSAJXTE-CLONMANBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neoxanthin belongs to the class of carotenoids. It is an allenic xanthophyll and a precursor of the plant hormone abscisic acid, which is formed as the last product of carotenoid synthesis in green plants.[1]

Application

Neoxanthin may be used as a reference standard in the determination of the analyte in fruits using high-performance liquid chromatography (HPLC) coupled with diode array detector (DAD) and mass spectrometry (MS).[1])

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of neoxanthin synthase as a carotenoid cyclase paralog.
Bouvier F, et al.
European Journal of Biochemistry, 267(21), 6346-6352 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service